Hand and finger protectors, Azlon®
Proveedor: SciLabware
PGR406
AZLOPGR406EA 56.3 EUR
AZLOPGR406
Hand and finger protectors, Azlon®
Protectores de manos
Hand protector made of silicone rubber.
  • Ideal for safe handling of hot beakers, flasks etc
  • Capable of withstanding temperatures up to 250 °C
  • Also excellent for handling cold objects down to −57 °C
  • Contact surfaces are multi-studded for positive grip
* One size fits all.