Azul brillante Coomassie® R-250 ~50% Dye, technical, Sigma-Aldrich®

Proveedor: SIGMA ALDRICH MICROSCOPY

Sinónimos: Hidrogeno(etil)[4-[4-[etil(m-sulfonatobencil)amino]-4'-(p-etoxianilino)bencidrilen]ciclohexa-2,5-dien-1-iliden](m-sulfonatobencil)amonio sal de monosodio

MQ200

B0149-100G B0149-25G B0149-500G B0149-5G
B0149-100GEA 256 EUR
B0149-100G B0149-25G B0149-500G B0149-5G
Azul brillante Coomassie® R-250 ~50% Dye, technical, Sigma-Aldrich®
Azul brillante Coomassie® R-250

Brilliant Blue R (C.l. 42660) is a triaminotriarylmethane dye with two amino groups being benzylated and one phenylated. It is also known as brilliant indocyanine 6B and coomassie blue R250. Brilliant Blue R has been widely used for protein band detection after SDS-PAGE (sodium dodecyl sulfate-polyacrylamide gel electrophoresis).


  • Its other applications include:
  • as a protein stain on epoxy-embedded tissue sections
  • for quantitative micro densitometry of protein in sections
  • for staining chromatids, actinin microfilaments, developing neurites, and sperm acrosomes
  • For detection of protein bands following electrophoresis

Formula: C₄₅H₄₄N₃NaO₇S₂
MW: 825.98 g/mol
Pto. de fusión: 174…180 °C
Temperatura de almacenaje: Room Temperature
Número MDL: MFCD00041762
Núm. CAS: 6104-59-2
EINECS: 228-060-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bccf790aff5db31beb2ef716ba4d09164ada98fd&vid=646beb4d76f324a18c02178d5530bfa89824a7a2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance (Form) Powder
Suitability For Detection of Protein Bands Following Electrophoresis Suitable
Solubility (Colour) Very Dark Purple to Very Dark Blue and Dark Blue-Black
Carbon 32.0 - 42.0 %
Nitrogen 2.5 - 3.5 %
Dye Content Approximately 50%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR