(S)-Hidrogenopirrolidina-1,2-dicarboxilato de 1-(9H-fluoren-9-ilmetilo) ≥98%

Proveedor: Thermo Fisher Scientific

B21081.06 B21081.14
B21081.06EA 53.6 EUR
B21081.06 B21081.14
(S)-Hidrogenopirrolidina-1,2-dicarboxilato de 1-(9H-fluoren-9-ilmetilo) ≥98%
(S)-Hidrogenopirrolidina-1,2-dicarboxilato de 1-(9H-fluoren-9-ilmetilo)

Purity: 98% CAS No.: 71989-31-6 Molecular Formula: C20H19NO4 Formula Weight: 337.38 Storage Temperature: 2°C to 8°C Melting Point: 113-117° Beilstein Reference: 3596735 Optical Rotation: -40° (c=1 in ethyl acetate) MDL No.: MFCD00037122

Formula: C₂₀H₁₉NO₄
Pto. de fusión: 113…117 °C
Número MDL: MFCD00037122
Núm. CAS: 71989-31-6
EINECS: 276-259-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR