Manganeso (II) acetato, anhidro 98+%

Proveedor: Thermo Fisher Scientific
Atención

B23708.0B B23708.18 B23708.30
B23708.0BEA 477 EUR
B23708.0B B23708.18 B23708.30
Manganeso (II) acetato, anhidro 98+%
Manganeso (II) acetato

Purity: 98+% CAS No.: 638-38-0 Molecular Formula: C4H6MnO4 Formula Weight: 173.03 Merck Reference: 14,5724 MDL No.: MFCD00013039

Formula: (H₃CCOO)₂Mn
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00013039
Núm. CAS: 638-38-0
EINECS: 211-334-3
Índice Merck: 14,5724

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR