DL-Methionine sulphone ≥98%

Proveedor: Thermo Fisher Scientific

B25094.03 B25094.06
B25094.03EA 41.3 EUR
B25094.03 B25094.06
DL-Methionine sulphone ≥98%
DL-Methionine sulphone

Purity: 98% CAS No.: 820-10-0 Molecular Formula: C5H11NO4S Formula Weight: 181.21 MDL No.: MFCD00025079

Formula: C₅H₁₁NO₄S
MW: 181.21 g/mol
Temperatura de almacenaje: Ambiente
Número MDL: MFCD00025079
Núm. CAS: 820-10-0
EINECS: 212-466-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR