Liquid nitrogen storage vessels, Thermo-Flask®
Proveedor: Thermo Fisher Scientific
2122DBQ 2123 2124 2129 2130
BARN2122DBQEA 510 EUR
BARN2122DBQ BARN2123 BARN2124 BARN2129 BARN2130
Liquid nitrogen storage vessels, Thermo-Flask®
Dewars y recipientes criogénicos
These liquid nitrogen storage vessels, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.
  • Durable, corrosion-resistant stainless-steel exterior
  • Ideal for use as a cold trap
  • 'Push-fit' insulated lid, vented to prevent pressure build-up
  • Stainless-steel jacket and cushioned base for years of trouble-free use
  • Rugged, shock-absorbing plastic exterior with carry handle
It is a convenient dish-shaped design perfect for benchtop use with crushed ice and dry ice/alcohol.