Digital heating/cooling block heater
Proveedor: Thermo Fisher Scientific
88880030
BARN88880030EA 1640 EUR
BARN88880030
Digital heating/cooling block heater
Termobloques Enfriadores / calentadores de bloque
Compact programmable heating/cooling block heater with digital display can be used for all common applications in laboratory. Applications include enzyme reactions, transformation, DNA extractions, reactions that need to processed at or near room temperature and warming and cooling of reagents.
  • PID control ensures consistent precise temperature control
  • Stores 10 programs and 10 steps per program
  • Cord set with various plug types
  • Low-profile design
  • Blocks easily interchanged
Información para pedidos: Blocks must be ordered separately.