Thermo Scientific™ Digital Mini Rotator
Proveedor: Thermo Fisher Scientific
88882008
BARN88882008EA 1340 EUR
BARN88882008
Thermo Scientific™ Digital Mini Rotator
Agitadores de laboratorio Agitadores Basculantes y de Rotación
Provides smooth, accurate and orbital mixing of various vessels in the laboratory.
  • Precise speed control: ±1 min⁻¹ (≤100 min⁻¹), ±1% (101 to 450 min⁻¹)
  • Orbit diameter: 10 mm
  • 2 timer modes: Continuous, Timer
  • Multi-platform choices for loading all sizes vessels in the laboratory