Arctic Express™ Cryogenic Shippers with Datalogger
Proveedor: Thermo Fisher Scientific
CY50905D CY50910D
BARNCY50905DEA 5110 EUR
BARNCY50905D BARNCY50910D
Arctic Express™ Cryogenic Shippers with Datalogger
Dewars y recipientes criogénicos
Thermo Scientific™ Arctic Express™ cryogenic shippers are designed to ensure the maintenance of sample integrity when movement of your samples is required. Arctic Express shippers come available with a built-in 21CFR, part 11 compliant datalogger, providing you with additional peace of mind that your important samples have been maintained during transport.
  • Innovative material absorbs liquid nitrogen to prevent spillage during shipment
  • 14- and 21-day liquid nitrogen holding times allow specimens to be shipped safely
  • Durable construction and sturdy base