Serum separator tubes, SST™ II advance, BD Vacutainer®
Proveedor: BD Medical
367957 367955 367953 367954 367956 367958 368879 366882 368969 368965 368966 368970
BDAM367957EA 54.2 EUR
BDAM367957 BDAM367955 BDAM367953 BDAM367954 BDAM367956 BDAM367958 BDAM368879 BDAM366882 BDAM368969 BDAM368965 BDAM368966 BDAM368970
Serum separator tubes, SST™ II advance, BD Vacutainer®
Sistemas para diagnóstico Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, clear.
  • Silica (Clot Activator)/Gel
  • Stable barrier between serum and clotted blood, therefore better analyte stability
  • Better sample quality
  • Optimisation of the work flow: Short centrifugation time, sample processing and archiving in the primary tube
  • No possibility of misidentification due to the use of secondary tubes
During the centrifugation of the BD Vacutainer® SST™ II Advance tubes, an inert gel separates the serum and the blood clot preventing the contamination of the serum from the separated cellular components. For example, the serum forMore Product Information