Stopcock, three way, BD Connecta™
Proveedor: BD Medical
394501
BDAM394501EA 289 EUR
BDAM394501
Stopcock, three way, BD Connecta™
Válvulas de cierre
This three way stopcock has a flow rate of 23,4 L/hr and a prime volume of 0,31 ml.