Distillation apparatus
Proveedor: BEHR
B00218232 B00218234 B00218039
BEHRB00218232EA 728 EUR
BEHRB00218232 BEHRB00218234 BEHRB00218039
Distillation apparatus
Destiladores Sistemas de destilación automática
Distillation apparatus for determining the alcohol content and the volatile acids in wine and other alcoholic drinks.
  • Suitable for high sample volumes
  • User can save the data and print them out
The supplied Windows software allows a bidirectional transmission of application-specific distillation parameters using the RS232 interface between one or more devices and a PC. A library with current applications is already included on the CD. The RS232 interface also allows data to be transmitted from the device to the PC during operation. The user can saveMore Product Information