Racks for cryogenic vial boxes, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F188530004
BELAF188530004EA 177 EUR
BELAF188530004
Racks for cryogenic vial boxes, SP Bel-Art
Gradillas Soportes de criocajas
Cryo storage towers are ideal for benchtop, incubator use, transportation, freezer and general purpose storage.
  • PC
Footprint dimensions: 150×150 mm
Shelf dimensions: 142×142 mm