Test tube racks, Stack Rack™, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F188601316 F188602630
BELAF188601316EA 82.9 EUR
BELAF188601316 BELAF188602630
Test tube racks, Stack Rack™, SP Bel-Art
Gradillas Tube Racks
PP, white
  • Sturdy and chemical resistant
  • Will not shatter or stain
  • With top shelf height of 70 mm
These rigid racks features interlocking end plates with handles for secure stacking when loaded with tubes up to 100 mm long. Rows are numbered and lettered for easy tube identification.