Racks for centrifuge bottles, Poxygrid®, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F198560500
BELAF198560500EA 73.5 EUR
BELAF198560500
Racks for centrifuge bottles, Poxygrid®, SP Bel-Art
Gradillas Tube Racks
This unique steel wire rack allows storage and transport of up to 4×500 ml conical bottom centrifuge tubes.
  • Autoclavable
  • Temperature range –223 to 121 °C
  • Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis