Sample spoons, mini, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
F367210005 F367210010 F367210050 F367210025 F367210100
BELAF367210005EA 54.6 EUR
BELAF367210005 BELAF367210010 BELAF367210050 BELAF367210025 BELAF367210100
Sample spoons, mini, SP Bel-Art
Cucharas
PS, white.
  • Disposable, made from white high-impact PS
  • Ideal for collecting specific small volumes of powders and other dry materials
  • 0,10 / 0,50 / 1,0 ml models have a rounded bowl for easier emptying of the spoon and a thinner neck that minimises extra material collection