Pipette stand, Poxygrid®, SP Bel-Art
Proveedor: Bel-Art Products, a Part of SP
H189620006
BELAH189620006EA 204 EUR
BELAH189620006
Pipette stand, Poxygrid®, SP Bel-Art
Soportes de pipetas
Pipette stand made of stainless steel.
  • Uncoated stainless steel is corrosion resistant and easy to clean
  • Supports 5 pipettors
  • Suitable for most popular brands including Gilson Pipetman® pipettor