Dropping funnels, cylindrical, without pressure balance
Proveedor: SciLabware
443333 443340 443347 443354 443360
BIBB443333EA 130 EUR
BIBB443333 BIBB443340 BIBB443347 BIBB443354 BIBB443360
Dropping funnels, cylindrical, without pressure balance
Embudos Embudos de goteo
Dropping funnel, cylindrical shape, made of Pyrex® borosilicate glass, with cone and socket. Stopcock with interchangeable PTFE key.
  • Excellent chemical and temperature resistance
  • White enamel double graduated scale
  • Stem inside cone for easy fitting to Quickfit® assemblies
Certificaciones: Complies with ISO 4800.