High purity hardened qualitative filter paper
Proveedor: Ahlstrom-Munksjö
113170 113123 113053 113041 113045 113048 113057 113060 113063 113068 113115 113116 113118 113119 113121 113124 113127 113165 113166 113168 113172 113175 113178 113180 113210 113211 113212 113213 113214 113215 113216 113217 113243 113244 113245 113246 113247 113248 113249 113250 113076 113078 113080 113083 113085 113087 113089 113136 113137 113140 113141 113146 113147 113148 113185 113187 113189 113191 113194 113195 113196 113221 113223 113225 113228 113230 113231 113233 113254 113255 113257 113258 113259 113260 113262 113022 113105 113205 113238 113039 113135
BINZ113170EA 18.8 EUR
BINZ113170 BINZ113123 BINZ113053 BINZ113041 BINZ113045 BINZ113048 BINZ113057 BINZ113060 BINZ113063 BINZ113068 BINZ113115 BINZ113116 BINZ113118 BINZ113119 BINZ113121 BINZ113124 BINZ113127 BINZ113165 BINZ113166 BINZ113168 BINZ113172 BINZ113175 BINZ113178 BINZ113180 BINZ113210 BINZ113211 BINZ113212 BINZ113213 BINZ113214 BINZ113215 BINZ113216 BINZ113217 BINZ113243 BINZ113244 BINZ113245 BINZ113246 BINZ113247 BINZ113248 BINZ113249 BINZ113250 BINZ113076 BINZ113078 BINZ113080 BINZ113083 BINZ113085 BINZ113087 BINZ113089 BINZ113136 BINZ113137 BINZ113140 BINZ113141 BINZ113146 BINZ113147 BINZ113148 BINZ113185 BINZ113187 BINZ113189 BINZ113191 BINZ113194 BINZ113195 BINZ113196 BINZ113221 BINZ113223 BINZ113225 BINZ113228 BINZ113230 BINZ113231 BINZ113233 BINZ113254 BINZ113255 BINZ113257 BINZ113258 BINZ113259 BINZ113260 BINZ113262 BINZ113022 BINZ113105 BINZ113205 BINZ113238 BINZ113039 BINZ113135
High purity hardened qualitative filter paper
Filtros Papeles de filtros Papel de filtro cualitativo
Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened gradesMore Product Information