High purity qualitative filter paper
Proveedor: Ahlstrom-Munksjö
110125 113334 113336 113338 113341 113344 113346 113348 113425 110100 113415 113414 113304 113314 113368 113411 113410 113409 113407 113404 113377 113376 113306 113370 113366 113364 113362 113327 113323 113317 110109 110111 110112 110113 110010 110098 110101 110122 110123 110124 110127 110128 113382 113384 113388 113389 113390 113391 113417 113421 113423 113427 113428
BINZ110125EA 28.4 EUR
BINZ110125 BINZ113334 BINZ113336 BINZ113338 BINZ113341 BINZ113344 BINZ113346 BINZ113348 BINZ113425 BINZ110100 BINZ113415 BINZ113414 BINZ113304 BINZ113314 BINZ113368 BINZ113411 BINZ113410 BINZ113409 BINZ113407 BINZ113404 BINZ113377 BINZ113376 BINZ113306 BINZ113370 BINZ113366 BINZ113364 BINZ113362 BINZ113327 BINZ113323 BINZ113317 BINZ110109 BINZ110111 BINZ110112 BINZ110113 BINZ110010 BINZ110098 BINZ110101 BINZ110122 BINZ110123 BINZ110124 BINZ110127 BINZ110128 BINZ113382 BINZ113384 BINZ113388 BINZ113389 BINZ113390 BINZ113391 BINZ113417 BINZ113421 BINZ113423 BINZ113427 BINZ113428
High purity qualitative filter paper
Filtros Papeles de filtros Papel de filtro cualitativo
These qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers cover a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened grades contain aMore Product Information