General purpose qualitative filter papers
Proveedor: Ahlstrom-Munksjö
123120 120014 120015 120016 120017 120018 120019 120021 120065 120012 120013 120004 120006 120007 120008 120010 120000 120002 120003 120032 120033 120034 120035 120036 120037 120040 120029 120030 120031 120038 123255 123256 123250 123251 123253 123260 123263 123011 123008 123010 123012 123151 123169 123171 123172 123242 123002 123145 123471 123321 123073 123091 123316 123007 123117 123334 123051 123299 123437 123084 123109 123107 123100 123098 123095 123093 123113 123087 123080 123074 123019 123018 123016 123015 123013 123315 123089 123313 123312 123111 123208 123176 123175 123174 123170 123006 123317 123314 123344 123332 123331 123330 123329 123328 123327 123323 123318 123584 123476 123478 123268 123261 123264 123249 123257 123254 123474 123269 123366 120055 123021 123022 123023 123024 123025 123026 123465 123466 123467 123554 123555 123558
BINZ123120EA 66.7 EUR
BINZ123120 512-6106 512-6105 512-6104 512-6103 512-6102 512-6101 512-6100 512-6109 512-6108 512-6107 512-6096 512-6094 512-6093 512-6092 512-6090 512-6099 512-6098 512-6097 512-6117 512-6115 512-6114 512-6113 512-6112 512-6111 512-6110 512-6121 512-6120 512-6119 512-6193 512-6195 512-6196 512-6235 512-6236 512-6238 512-6245 512-6247 516-0750 516-0748 516-0749 516-0751 516-0757 BINZ123169 BINZ123171 BINZ123172 BINZ123242 BINZ123002 BINZ123145 BINZ123471 BINZ123321 BINZ123073 BINZ123091 BINZ123316 516-0747 BINZ123117 BINZ123334 BINZ123051 BINZ123299 BINZ123437 BINZ123084 BINZ123109 BINZ123107 BINZ123100 BINZ123098 BINZ123095 BINZ123093 BINZ123113 BINZ123087 BINZ123080 BINZ123074 516-0756 516-0755 516-0754 516-0753 516-0752 BINZ123315 BINZ123089 BINZ123313 BINZ123312 BINZ123111 BINZ123208 BINZ123176 BINZ123175 BINZ123174 BINZ123170 516-0746 BINZ123317 BINZ123314 BINZ123344 512-0606 BINZ123331 BINZ123330 BINZ123329 BINZ123328 BINZ123327 BINZ123323 BINZ123318 BINZ123584 BINZ123476 BINZ123478 BINZ123268 516-0770 516-0771 516-0767 516-0769 516-0768 BINZ123474 BINZ123269 BINZ123366 BINZ120055 BINZ123021 BINZ123022 BINZ123023 BINZ123024 BINZ123025 BINZ123026 BINZ123465 BINZ123466 BINZ123467 BINZ123554 BINZ123555 BINZ123558
General purpose qualitative filter papers
Filtros Papeles de filtros Papel de filtro cualitativo
Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Standard qualitative filterMore Product Information