Filter papers for soil analysis
Proveedor: Ahlstrom-Munksjö
146032 146030 146029 146020
BINZ146032EA 67.1 EUR
BINZ146032 BINZ146030 BINZ146029 516-0776
Filter papers for soil analysis
Filtros Papeles de filtros Papel de filtro cualitativo
These papers are used to determine the presence of trace elements and nutrients in soil samples to optimise plant growth.
Order Now
ESPECIFICACIONES
Grado 132
Environmentally Preferable
Peso 80 g/m²
Velocidad de filtración 450 s (Herzberg)