Glass microfibre filters for air particle monitoring, grade MGR and MG 10
Proveedor: Ahlstrom-Munksjö
410336 410337 410350 410354 410357 410335 410315 410318 410319 410320 410321 410324 410325 410326 410327 410328 410329 410330 410331 410334 410338 410340 410341 410343 410349 410351 410352 410353 410355 410356
BINZ410336EA 144 EUR
BINZ410336 BINZ410337 BINZ410350 BINZ410354 BINZ410357 BINZ410335 BINZ410315 BINZ410318 BINZ410319 BINZ410320 BINZ410321 BINZ410324 BINZ410325 BINZ410326 BINZ410327 BINZ410328 BINZ410329 BINZ410330 BINZ410331 BINZ410334 BINZ410338 BINZ410340 BINZ410341 BINZ410343 BINZ410349 BINZ410351 BINZ410352 BINZ410353 BINZ410355 BINZ410356
Glass microfibre filters for air particle monitoring, grade MGR and MG 10
Filtros Filtros de fibras de vidrio
These Grade MGR and MG 10 grade filters are hydrophobic micro-glass fibre filters with binder suitable for use when low affinity with water and moisture are required. These high-quality materials are designed for the specific needs of air monitoring analyses.
  • Air sampling for the collection of atmospheric particulates and aerosols
  • Particle filtration of gasses
  • Air filter media
  • Very high-water repellency and strong resistance to tearing and breaking, while maintaining a low thickness
MGR is a very thin micro-glass hydrophobic material designedMore Product Information