Anti-ETV7 Rabbit polyclonal antibody

Proveedor: Biorbyt
ORB101078-100 ORB101078-200
BIRBORB101078-100EA 0 EUR
BIRBORB101078-100 BIRBORB101078-200
Anti-ETV7 Rabbit polyclonal antibody
Anticuerpos
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: ETV7 (ets variant 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ETV7
Environmentally Preferable
Nombre del antígeno ETS variant 7
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Rabbit
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
Forma liquid
Molecular weight 39 kDa
Purification purified by affinity chromatography
Concentración 0,5 mg/ml
Storage buffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR