Anti-IRF7 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB108971-100
BIRBORB108971-100EA 0 EUR
BIRBORB108971-100
Anti-IRF7 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: IRF7 (interferon regulatory factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR