Anti-STK24 Mouse Polyclonal Antibody

Proveedor: Biorbyt
ORB11347-100 ORB11347-200
BIRBORB11347-100EA 0 EUR
BIRBORB11347-100 BIRBORB11347-200
Anti-STK24 Mouse Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: STK24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR