Anti-MAP3K1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128315-100 ORB128315-200
BIRBORB128315-100EA 0 EUR
BIRBORB128315-100 BIRBORB128315-200
Anti-MAP3K1 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: MAP3K1 (mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno MAP3K1
Nombre del antígeno Mitogen-activated protein kinase kinase kinase 1, E3 ubiquitin Protein ligase
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human,Rat,Mouse
Huésped Rabbit
Isotipo
Western blot Yes
InmunoQuímica Yes
Modificación phospho-Thr1402
Purification purified by affinity chromatography
Concentración 1 mg/ml
Storage buffer PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR