Anti-CKAP4 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB128863-100 ORB128863-200
BIRBORB128863-100EA 0 EUR
BIRBORB128863-100 BIRBORB128863-200
Anti-CKAP4 Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CKAP4 (cytoskeleton-associated protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR