Anti-ADH Rabbit Polyclonal Antibody (Biotin)

Proveedor: Biorbyt
ORB21035-1
BIRBORB21035-1EA 0 EUR
BIRBORB21035-1
Anti-ADH Rabbit Polyclonal Antibody (Biotin)
Anticuerpos
Type: Primary
Antigen: ADH (alcohol dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR