Anti-AGFG1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB2313-100 ORB2313-200
BIRBORB2313-100EA 0 EUR
BIRBORB2313-100 BIRBORB2313-200
Anti-AGFG1 Rabbit Polyclonal Antibody
Anticuerpos
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: AGFG1 (ArfGAP with FG repeats 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR