Anti-ACTB Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB74081-100
BIRBORB74081-100EA 0 EUR
BIRBORB74081-100
Anti-ACTB Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: ACTB (actin, beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR