Anti-S1PR3 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB85350-50
BIRBORB85350-50EA 0 EUR
BIRBORB85350-50
Anti-S1PR3 Rabbit Polyclonal Antibody
Anticuerpos
Immunochemistry: 6 ?g/ml

Type: Primary
Antigen: S1PR3 (sphingosine-1-phosphate receptor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR