Anti-TRDMT1 Mouse Monoclonal Antibody

Proveedor: Biorbyt
ORB87974-50
BIRBORB87974-50EA 0 EUR
BIRBORB87974-50
Anti-TRDMT1 Mouse Monoclonal Antibody
Anticuerpos
Immunochemistry: 5 ?g/ml; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: TRDMT1 (tRNA aspartic acid methyltransferase 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR