Anti-NOD2 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB88404-50
BIRBORB88404-50EA 0 EUR
BIRBORB88404-50
Anti-NOD2 Rabbit Polyclonal Antibody
Anticuerpos
Immunofluorescence: 10 ?g/ml; Immunochemistry: 3 ?g/ml; Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: NOD2 (nucleotide-binding oligomerization domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR