Anti-MTA1 Goat Polyclonal Antibody

Proveedor: Biorbyt
ORB89694-50
BIRBORB89694-50EA 0 EUR
BIRBORB89694-50
Anti-MTA1 Goat Polyclonal Antibody
Anticuerpos
ELISA: 1:16000; Immunochemistry: 3.75 ?g/ml; Western Blot: 0.03-0.1 ?g/ml

Type: Primary
Antigen: MTA1 (metastasis associated 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno MTA1
Nombre del antígeno Metastasis associated 1
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Unconjugated
Reactividad Human
Huésped Goat
Isotipo
Western blot Yes
ELISA Yes
InmunoQuímica Yes
Purification purified by immunoaffinity chromatography
Storage buffer Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
Temperatura de almacenamiento Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR