Anti-GRTP1 Rabbit Polyclonal Antibody

Proveedor: Biorbyt
ORB94256-50
BIRBORB94256-50EA 0 EUR
BIRBORB94256-50
Anti-GRTP1 Rabbit Polyclonal Antibody
Anticuerpos
Immunochemistry: 5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: GRTP1 (growth hormone regulated TBC protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR