Anti-MAG Mouse Monoclonal Antibody

Proveedor: Biorbyt
ORB95109-50
BIRBORB95109-50EA 0 EUR
BIRBORB95109-50
Anti-MAG Mouse Monoclonal Antibody
Anticuerpos
Immunochemistry: 2.5 ?g/ml; Western Blot: 1:500-1:1000

Type: Primary
Antigen: MAG (myelin associated glycoprotein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR