Anti-TFF1 Mouse Monoclonal Antibody (CF568) [clone: GE2]

Proveedor: Biotium
BNC680677-500 BNC680677-100
BNC680677-500EA 560 EUR
BNC680677-500 BNC680677-100
Anti-TFF1 Mouse Monoclonal Antibody (CF568) [clone: GE2]
Anticuerpos
This antibody recognizes a polypeptide of 6.5 kDa, identified as pS2 estrogen-regulated protein. Its epitope is localized between aa57-84 of human pS2 protein. pS2 is a trefoil peptide. Trefoil peptides are protease resistant molecules secreted throughout the gut that play a role in mucosal healing. These peptides contain three intra-chain disulfide bonds, forming the trefoil motif, or P-domain. pS2 is known to form dimers and this dimerization is thought to play a role in its protective and healing properties. About 60% of breast carcinomas are positive for pS2. Staining is cytoplasmic,More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno TFF1 TFF1
Environmentally Preferable
Nombre del antígeno pS2 pS2
Sinónimos de antígenos Trefoil Factor 1|Gastrointestinal trefoil protein pS2|HPS2|BCEI|Gastrointestinal Trefoil Protein|HP1A|TFF1|pNR2|Breast Cancer Estrogen Inducible Protein Trefoil Factor 1|Gastrointestinal trefoil protein pS2|HPS2|BCEI|Gastrointestinal Trefoil Protein|HP1A|TFF1|pNR2|Breast Cancer Estrogen Inducible Protein
Tipo de anticuerpo Primario Primario
Clonalidad Monoclonal Monoclonal
Conjugado CF® 568 CF® 568
Clon Clone GE2 Clone GE2
Reactividad Cynomolgus monkey,Human Cynomolgus monkey,Human
Huésped Mouse Mouse
ID del gen 162807 162807
Isotipo IgG1 kappa IgG1 kappa
InmunoQuímica Yes Yes
FluorescenciaInmunológica Yes
Concentración 0,1 mg/ml 0,1 mg/ml
Storage buffer PBS/0,1% BSA/0,05% azide PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR