Anti-KRT8 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: H1]
Proveedor: Biotium
BNUM0334-50
BNUM0334-50EA 417 EUR
BNUM0334-50
Anti-KRT8 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: H1]
Anticuerpos
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). CK8 is primarily found in the non-squamous epithelia and is present in majority of adenocarcinomas and ductal carcinomas. It is absent in squamous cell carcinomas. Hepatocellular carcinomas are defined by the use of antibodies that recognize only cytokeratin 8 and 18. CK8 exists on several types of normal and neoplastic epithelia, including many ductal and glandular epithelia such as colon, stomach, small intestine, trachea, andMore Product Information