Anti-AFP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: C2]

Proveedor: Biotium
BNUM0523-50
BNUM0523-50EA 417 EUR
BNUM0523-50
Anti-AFP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: C2]
Anticuerpos
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP) (ISOBM TD-2 workshop). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. AFP is normally synthesized in the liver, intestinal tract, and yolk sac of the fetus. Antibody to AFP has been shown to be useful in detecting hepatocellular carcinomas (HCC) and germ cell neoplasms, especially yolk sac tumors.

Type: Primary
Antigen: AFP
Clonality: Monoclonal
Clone: C2
Conjugation: Purified, BSA-free
Epitope:
Host:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR