Anti-ITGB3 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 2.50E 12]

Proveedor: Biotium
BNUM1048-50
BNUM1048-50EA 417 EUR
BNUM1048-50
Anti-ITGB3 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 2.50E 12]
Anticuerpos
Reacts with human integrin beta3 (GPIIIa, vitronectin receptor beta chain). The protein detectable is a complex of CD41 and CD61.The apparent molecular weight of the GPIIIa by SDS-PAGE is 105 kDa reduced and 90 kDa unreduced. Ligands are fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin. Residues 237-248 of GPIIIa or CD61 are critical in adhesive protein binding.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: 2.50E 12
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno ITGB3
Nombre del antígeno CD61
Environmentally Preferable
Sinónimos de antígenos HPA 1|Platelet fibrinogen receptor beta subunit|ITGB3|PTP|NAIT|Platelet glycoprotein IIIa|GP3A|BDPLT2|Integrin beta-3|HPA 4|GPIIIa|GT
Tipo de anticuerpo Primario
Clonalidad Monoclonal
Conjugado Unconjugated
Clon Clone 2.50E 12
Reactividad Human
Huésped Mouse
ID del gen 218040
Isotipo IgM kappa
FluorescenciaInmunológica Yes
Purification BSA-free
Concentración 1 mg/ml
Storage buffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR