Retort clamp, MAXI
Proveedor: BOCHEM
5567
BOCH5567EA 305 EUR
BOCH5567
Retort clamp, MAXI
Abrazaderas Pinzas para uso general
Retort clamp made of chromium plated steel.
  • Fibreglass covered
  • 400 mm total length