Overhead stirrer tools, centrifugal stirrers
Proveedor: BOCHEM
9210 9211 9212 9213 9214 9209 9215 9216
BOCH9210EA 27.6 EUR
BOCH9210 BOCH9211 BOCH9212 BOCH9213 BOCH9214 441-1905 BOCH9215 BOCH9216
Overhead stirrer tools, centrifugal stirrers
Imánes de agitación Herramientas de agitación
Centrifugal stirrers made of stainless steel, with two flexible blades.
  • Ideal for stirring liquids at middle speed range
  • Suitable for stirring low to medium viscous media
  • Creates radial flow
  • Suitable for jars with tight neck