BOD calibration standards, (CRM), Supelco®
Proveedor: Merck
BOD1000-500ML BOD2000-500ML BOD200-100ML BOD200-500ML BOD500-500ML
BOD1000-500MLEA 127 EUR
BOD1000-500ML BOD2000-500ML BOD200-100ML BOD200-500ML BOD500-500ML
BOD calibration standards, (CRM), Supelco®
Normativas Estándar par análisis medioambiental
Certificaciones: These certified reference materials (CRM) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Precaución: Storage temperature: 2 - 8 °C (refrigerator)