Tube rotator
Proveedor: BOEKEL SCIENTIFIC
260650
BOEI260650EA 1370 EUR
BOEI260650
Tube rotator
Agitadores de laboratorio Agitadores Basculantes y de Rotación
Five carousel options with adjustable speed and tilt angles provide greater versatility.
Timer feature offers time down or time up counting. Digital microprocessor controlled. Supplied with a 14×50 ml tube carousel. 2-year warranty.