Magnetic stirring bars, cylindrical, PTFE, BOLA
Proveedor: Bohlender
C350-03 C350-04 C350-08 C350-13 C350-22 C350-24 C350-41 C350-26 C350-37 C350-31 C350-35 C350-36 C350-16 C350-18 C350-07 C350-27 C350-25 C350-23 C350-21 C350-20 C350-19 C350-17 C350-15 C350-14 C350-12 C350-11 C350-29 C350-09 C350-30 C350-06 C350-05 C350-02 C350-01 C350-10 C350-39 C350-34 C350-33 C350-32
BOHLC350-03EA 1.7 EUR
BOHLC350-03 BOHLC350-04 BOHLC350-08 BOHLC350-13 BOHLC350-22 BOHLC350-24 442-9840 442-9841 442-9842 BOHLC350-31 BOHLC350-35 BOHLC350-36 BOHLC350-16 BOHLC350-18 BOHLC350-07 BOHLC350-27 BOHLC350-25 BOHLC350-23 BOHLC350-21 BOHLC350-20 BOHLC350-19 BOHLC350-17 BOHLC350-15 BOHLC350-14 BOHLC350-12 BOHLC350-11 BOHLC350-29 BOHLC350-09 BOHLC350-30 BOHLC350-06 BOHLC350-05 BOHLC350-02 BOHLC350-01 BOHLC350-10 BOHLC350-39 BOHLC350-34 BOHLC350-33 BOHLC350-32
Magnetic stirring bars, cylindrical, PTFE, BOLA
Imánes de agitación Barras magnéticas de agitación
Standard magnetic stirring bar with PTFE-encapsulated magnetic core (Alnico 5).
  • Universal chemical resistance
  • Temperature resistance from –200 to +250 °C
Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics.