Bellow connectors
Proveedor: Bohlender
H906-02 H906-04 H906-06 H906-12 H906-14 H907-10 H907-15
BOHLH906-02EA 60 EUR
BOHLH906-02 BOHLH906-04 BOHLH906-06 BOHLH906-12 BOHLH906-14 BOHLH907-10 BOHLH907-15
Bellow connectors
Adaptadores Adaptadores con juntas NS
Bellow connectors, made of PTFE, with sharp folds and sealing rings on the outside of the cone.
  • Universal chemical resistance
  • Suitable for vacuum applications
  • Temperature resistance from −200 to +250 °C
  • Compensates vibrations from vacuum pumps
  • Suiltable for length compensation of heated columns and angular misalignment