Waste Tube Connection, b.safe
Proveedor: Bohlender
R1105-25
BOHLR1105-25EA 200 EUR
BOHLR1105-25
Waste Tube Connection, b.safe
Tuberías Tubo de transferencia
For safe connection of canisters in safety cabinets to a waste disposal system with b.safe Waste Tubes.
  • Made of static dissipative PE-EX
  • A suitable attachment for fire protection is available separately
Mount for safety cabinets with a wall thickness of max. 100 mm and bore Ø 28 mm, lock nut with silicone gaskets for clamp fastening, 2 connections GL 25 male for connection to b.safe Waste Tubes.