Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Proveedor: Bioss
BS-0007R-A647
BOSSBS-0007R-A647EA 893 EUR
BOSSBS-0007R-A647
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticuerpos
AM and PAMP are potent hypotensive and vasodilatator agents. Numerous actions have been reported most related to the physiologic control of fluid and electrolyte homeostasis. In the kidney, am is diuretic and natriuretic, and both am and pamp inhibit aldosterone secretion by direct adrenal actions. In pituitary gland, both peptides at physiologically relevant doses inhibit basal ACTH secretion. Both peptides appear to act in brain and pituitary gland to facilitate the loss of plasma volume, actions which complement their hypotensive effects in blood vessels.

Recommended Dilutions: IF(IHC-More Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno ADM
Environmentally Preferable
Nombre del antígeno Adrenomedullin
Sinónimos de antígenos AM|Proadrenomedullin N-20 terminal peptide|ADM|PAMP|ProAM N-terminal 20 peptide|ProAM-N20
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 647
Reactividad Human,Rat
Huésped Rabbit
ID del gen 133
Isotipo IgG
InmunoQuímica Yes
Inmunógeno 115-146/185
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR