Anti-CA125 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Proveedor: Bioss
BS-0091R-A680
BOSSBS-0091R-A680EA 899 EUR
BOSSBS-0091R-A680
Anti-CA125 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces.

Type: Primary
Antigen: CA125
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 250-350/2008
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR